Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABH1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB105042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105042 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105042 Supplier Novus Biologicals Supplier No. NBP298658G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABH1 Polyclonal specifically detects ABH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ABH1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ABH1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 8846
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.