Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABH1 Antibody (OTI4C4), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271390B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271390B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271390B Supplier Novus Biologicals Supplier No. NBP271390B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ABH1 Monoclonal specifically detects ABH1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen ABH1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI4C4
Conjugate Biotin
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias ABH1, ABHalkB, alkylation repair homolog (E. coli), alkB, alkB, alkylation repair homolog 1 (E. coli), ALKBH, alkylated DNA repair protein alkB homolog 1, alkylation repair, alkB homolog, Alpha-ketoglutarate-dependent dioxygenase ABH1, DNA lyase ABH1, EC 1.14.11.-, EC 4.2.99.18, hABH
Gene Symbols ALKBH1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-245 of human ABH1 (NP_006011) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 8846.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.