Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABHD14B Antibody (011), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB038338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB038338 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB038338 Supplier Novus Biologicals Supplier No. NBP290260DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ABHD14B Monoclonal specifically detects ABHD14B in Human samples. It is validated for Western Blot.

Specifications

Antigen ABHD14B
Applications Western Blot
Classification Monoclonal
Clone 11
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias abhydrolase domain containing 14B, abhydrolase domain-containing protein 14B, CCG1-interacting factor B, CIBcell cycle gene 1-interacting factor B, EC 3.-, EC 3.1.11.6, EC 3.1.21.4, MGC15429
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human ABHD14B (Q96IU4-1; Met1-Gln210).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 84836
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.