Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abhd5 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB339311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339311 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339311 Supplier Novus Biologicals Supplier No. NB11041576PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Abhd5 Polyclonal antibody specifically detects Abhd5 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot

Specifications

Antigen Abhd5
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Simple Western
Formulation PBS
Gene Alias abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region within residues 200-300 of the human Abhd5 protein. [Swiss-Prot# Q8WTS1]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51099
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.