Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABHD7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $459.50
Specifications
Antigen | ABHD7 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145426
|
Invitrogen™
PA5145426 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145425
|
Invitrogen™
PA5145425 |
200 μL |
Each for $459.50
|
|
|||||
Description
ABHD7 gene ontology (GO) annotation includes hydrolase activity and membrane.Specifications
ABHD7 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q6IE26, Q8IUS5 | |
253152, 384214 | |
Recombinant Mouse EPHX4 protein, Lys36-Asp359 (Accession #Q6IE26). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
ABHD7; abhydrolase domain containing 7; abhydrolase domain-containing protein 7; EH4; EPHX4; Ephxrp; epoxide hydrolase 4; Epoxide hydrolase-related protein; Gm1382 | |
EPHX4 | |
IgG | |
Protein A | |
EPHX4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title