Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABI2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB084890 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB084890 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB084890 Supplier Novus Biologicals Supplier No. NBP299345C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABI2 Polyclonal specifically detects ABI2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen ABI2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias Abelson interactor 2, Abi-2, ABI2B, abl binding protein 3, abl interactor 2, Abl-binding protein 3, AblBP3ABI-2, abl-interacting protein 1 (SH3-containing protein), abl-interactor 2, abl-interactor protein 2b, AIP-1, arg protein tyrosine kinase-binding protein, Arg-binding protein 1, argBP1, ARGBPIA, argBPIB, SSH3BP2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human ABI2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10152
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.