Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABI3BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 277271AP150UL

Description
ABI3BP contains 2 fibronectin type-III domains. The loss of ABI3BP expression could play a functional role in thyroid tumorigenesis. It also presumably represents a trigger gene for evoking cellular senescence, which has also been suggested to be involved in the prevention of tumorigenesis.Specifications
ABI3BP | |
Polyclonal | |
Unconjugated | |
ABI3BP | |
ABI3BP, Nesh binding protein, NESHBP, Target of Nesh SH3, TARSH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25890, 320712 | |
-20°C | |
Liquid |
Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z7G0 | |
Abi3bp | |
ABI3BP Fusion Protein Ag26841 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction