Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ABI3BP Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ABI3BP |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5742
![]() ![]() |
Proteintech Group Inc
277271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
ABI3BP contains 2 fibronectin type-III domains. The loss of ABI3BP expression could play a functional role in thyroid tumorigenesis. It also presumably represents a trigger gene for evoking cellular senescence, which has also been suggested to be involved in the prevention of tumorigenesis.Specifications
ABI3BP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q7Z7G0 | |
25890, 320712 | |
ABI3BP Fusion Protein Ag26841 | |
Primary | |
-20°C | |
ABI3BP |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ABI3BP, Nesh binding protein, NESHBP, Target of Nesh SH3, TARSH | |
Abi3bp | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title