Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Akt Activity Assay Kit
Click to view available options
Quantity:
1 Kit
Description
- Akt is a protein kinase that can be activated by insulin and various growth factors and functions in a pathway involving PI3 kinase
- Recent evidence suggests that Akt functions to promote cell survival by actively inhibiting apoptosis
- Utilizes an Akt-specific antibody to immunoprecipitate Akt from cell lysate
- Kit specifically detects Akt1, Akt2, and Akt3 activities; other kinase activities would not be detected
Specifications
Specifications
Quantity | 1 Kit |
Description | Akt Activity Assay Kit is used for detecting Akt1, Akt2, and Akt3 activities. |
Content And Storage | Store the kit at -20°C |
For Use With (Application) | Functional Study, Immunoprecipitation, Western Blot |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction