Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Alkaline Phosphatase Assay Kit (Fluorometric)
Click to view available options
Sample Type:
Plasma, Serum
Description
Includes
- ALP assay buffer, 100mL
- MUP substrate, 1 vial
- ALP enzyme, 1 vial
- Stop solution, 25mL
Specifications
Specifications
Content And Storage | Store the kit at -20°C. |
Description | Alkaline Phosphatase Assay Kit is a quantitative fluorimetric determination of alkaline phosphatase at FL360/450 nm. |
Detection Method | Fluorometric |
For Use With (Application) | Functional Study |
Quantity | 1 Kit |
Sample Type | Plasma, Serum |
Sample Volume | 10 μL |
Type | Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction