Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Alkaline Phosphatase Assay Kitc
Click to view available options
Quantity:
1 Kit
Description
Includes
- ALP assay buffer, 100mL
- pNPP (10 TAB), 1 vial
- ALP enzyme, 1 vial
- Stop solution, 10mL
Specifications
Specifications
Calibration Range | 10 to 250μU |
Content And Storage | Store the kit at -20°C. |
Description | Alkaline Phosphatase Assay Kit is used for detecting ALP activity. |
Detection Method | Colorimetric |
For Use With (Application) | Functional Study |
Includes | 100mL ALP assay buffer, 1 vial pNPP (10 TAB), 1 vial ALP enzyme, 10mL stop solution |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
Type | Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction