Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ alpha-Ketoglutarate Assay Kit
Click to view available options
Quantity:
1 Kit
Description
Includes
- KG Assay Buffer 25mL -KG OxiRed Probe (in DMSO) 0.2mL
- KG Converting Enzyme (lyophilized) 1 vial
- KG Development Enzyme Mix (lyophilized) 1 vial
- KG Standard (10μmol, lyophilized) 1 vial
- ª-KG is an important nitrogen transporter
- One of the organic acids measured in newborns as an indicator of inborn errors of metabolism
- Kit provides a simple, sensitive, and rapid means for quantifying a-KG in a variety of samples
- Useful for detecting ª-ketoglutarate in the range of 0.01 to 10 nmoles
- Store kit at -20°C, protect from warm light
- Warm ª-KG Assay Buffer to room temperature before use
- Briefly centrifuge all small vials prior to opening
Specifications
Specifications
Quantity | 1 Kit |
Description | alpha-Ketoglutarate Assay Kit is used for quantifying alpha-KG. |
Content And Storage | Store the kit at -20°C |
Detection Method | Colorimetric, Fluorometric |
For Use With (Application) | Functional Study |
Calibration Range | 0.01 to 10 nm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction