Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Amylase Assay Kit
Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Content And Storage | Store the kit at -20°C. |
Description | Amylase Assay Kit is used for detecting amylase. |
Detection Method | Colorimetric |
For Use With (Application) | Functional Study |
Includes | 25mL Amylase assay buffer, 5mL Amylase substrate mix, 1 vial Amylase positive control (lyophilized), 150μL Nitrophenol standard (2mM) |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine, Food Sample, Fermentation Medium |
Type | Activity Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction