Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Annexin V-Cy3 Apoptosis Kit
Click to view available options
Quantity:
1 Kit
Description
- Kit is based on the observation that soon after initiating apoptosis, cells translocate the membrane phosphatidyl-serine (PS) from the inner face of the plasma membrane to the cell surface
- One-step staining procedure takes only 10 minutes
- Detection can be analyzed by flow cytometry or by fluorescence microscopy
- Store kit at 4°C; stable for one year
Specifications
Specifications
Description | Annexin V-Cy3 Apoptosis Kit is used for apoptosis detection. |
Quantity | 1 Kit |
Stability | 1 Year |
Content And Storage | Store the kit at 4°C. |
For Use With (Application) | Flow Cytometry, Functional Study |
Conjugate | Fluorescent |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction