Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Asparaginase Activity Assay Kit
Click to view available options
Quantity:
1 Kit
Description
Includes
- Asparaginase Assay Buffer 25mL WM
- OxiRed Probe (in DMSO) 0.2mL Red
- Substrate Mix 1 vial Orange
- Aspartate Enzyme Mix 1 vial Green
- Conversion Mix 1 vial Purple
- Positive Control 1 vial Blue
- Aspartate Standard (100mM) 0.1mL Yellow
Specifications
Specifications
Content And Storage | Store the kit at -20°C. For best results, store the components as described in the protocol. |
Description | Asparaginase Activity Assay Kit is a colorimetric and fluorometric method for assaying the activity of asparaginase. |
Detection Method | Colorimetric, Fluorometric |
For Use With (Application) | Functional Study |
Includes | 25mL Asparaginase assay buffer, 0.2mL OxiRed™ probe (in DMSO), 1 vial substrate mix (lyophilized), 1 vial Aspartate enzyme mix (lyophilized), 1 vial conversion mix (lyophilized), 1 vial positive control (lyophilized), 0.1mL Aspartate sta |
Quantity | 1 Kit |
Sample Type | Cell Lysate, Biological Sample, Food Sample |
Sample Volume | 50 μL |
Type | Activity Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction