Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Cytochrome c Apoptosis Detection Kit
Click to view available options
Quantity:
1 Kit
Description
- Provides an effective means for detecting cytochrome c translocation from mitochondria into cytosol during apoptosis
- Provides unique formulations of reagents to isolate a highly enriched mitochondria fraction from cytosol
- Procedure is simple and easy to perform; no ultracentrifugation is required and no toxic chemicals are involved
- Cytochrome c releasing from mitochondria into cytosol is then determined by Western blotting using the cytochrome c antibody provided in the kit
- Store at -20°C
Specifications
Specifications
Description | Cytochrome c Apoptosis Detection Kit is used for detecting cytochrome c translocation from mitochondria into cytosol during apoptosis. |
Quantity | 1 Kit |
Content And Storage | Store the kit at -20°C |
For Use With (Application) | Functional Study |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction