Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ DPP4 Activity Assay Kit (Fluorometric)
Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Content And Storage | Store the kit at -20°C. |
Description | DPP4 Activity Assay Kit is a quantitative fluorometric determination of DPP4 activity. |
Detection Method | Fluorometric |
For Use With (Application) | Functional Study |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Culture Supernatant, Fermentation Medium, Plasma, Serum, Tissue Supernatant, Urine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction