Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Fructose Assay Kit
Click to view available options
Quantity:
1 Kit
Description
- Sample Type: Culture Medium, Milk, Plasma, Saliva, Serum, Urine
- Sample Volume: 20μL
Functional Study
Specifications
Specifications
Calibration Range | 12 to 1000μM |
Content And Storage | Store the kit at -20°C. |
Description | Fructose Assay Kit is a quantitative colorimetric fructose determination at 565 nm. |
Detection Method | Colorimetric |
For Use With (Application) | Functional Study |
Includes | 10mL assay buffer, 120μL enzyme, 1.5mL PMS solution, 1.5mL MTT solution, 400μL standard 20mM D-Fructose |
Quantity | 1 Kit |
Sample Type | Culture Medium, Milk, Plasma, Saliva, Serum, Urine |
Sample Volume | 20 μL |
Type | Activity Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction