Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Glucose and Sucrose Assay Kit
Click to view available options
Quantity:
1 Kit
Specifications
Specifications
Content And Storage | Store the kit at -20°C. |
Description | Glucose and Sucrose Assay Kit is used for measuring glucose and sucrose levels. |
Detection Method | Colorimetric, Fluorometric |
For Use With (Application) | Functional Study |
Includes | 25mL Glucose assay buffer, 200μL Glucose probe (in DMSO), 1 vial Invertase (lyophilized), 1 vial Glucose enzyme mix (lyophilized), 100μL Sucrose standard (100nM/μL) |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
Type | Activity Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction