Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Glutamate Dehydrogenase Assay Kit
Shop All Abnova Corporation Products
Click to view available options
Sample Type:
Culture Medium, Plasma, Serum, Tissue, Urine
Description
Includes
- GDH assay buffer, 25mL
- Glutamate (2M), 1mL
- GDH developer (lyophilized), 1 vial
- GDH positive control (lyophilized), 1 vial
- NADH (0.5μmol, lyophilized), 1 vial
Specifications
Specifications
| Calibration Range | 0.4 to 80μ/L |
| Content And Storage | Store the kit at -20°C. |
| Description | Glutamate dehydrogenase Assay Kit is a quantitative colorimetric determination of glutamate dehydrogenase at 565 nm. |
| Detection Method | Colorimetric |
| For Use With (Application) | Functional Study |
| Quantity | 1 Kit |
| Sample Type | Culture Medium, Plasma, Serum, Tissue, Urine |
| Sample Volume | 20 μL |
| Type | Activity Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction