Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Glutathione Assay Kit
Click to view available options
Quantity:
1 Kit
Description
Includes
- Glutathione assay buffer, 30mL
- PCA (perchloric acid, 6N) 2mL
- KOH (6N) 2ml
- OPA probe (o-phthalaldehyde) 0.2mL
- Reducing agent mix, 1 vial
- GSH quencher, 20μL
- GSH standard (FW: 307), 1mg
Specifications
Specifications
Content And Storage | Store the kit at -20°C. |
Description | Glutathione Assay Kit is used for detecting GSH, GSSG, and total glutathione individually. |
Detection Method | Fluorometric |
For Use With (Application) | Functional Study |
Includes | 30mL Glutathione assay buffer, 2mL PCA (Perchloric Acid, 6N), 2mL KOH (6N), 0.2mL OPA probe (o-phthalaldehyde), 1 vial reducing agent mix, 20μL GSH quencher, 1mg GSH standard (FW: 307) |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
Type | Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction