Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Glutathione Peroxidase Assay Kit
Click to view available options
Quantity:
1 Kit
Description
- In this assay kit, GPx reduces Cumene Hydroperoxide while oxidizing GSH to GSSG
- Assay can be used to measure all of the glutathione dependent peroxidases in plasma, erythrocyte lysates, tissue homogenates, and cell lysates
- Kit has a detection sensitivity of approximately 0.5 mU/ml of GPx in samples
- Store kit at -20°C; protect from light
Specifications
Specifications
Quantity | 1 Kit |
Description | Glutathione Peroxidase Assay Kit is used for measuring glutathione peroxidase (GPx) activity. |
Content And Storage | Store the kit at -20°C |
Detection Method | Colorimetric |
For Use With (Application) | Functional Study |
Calibration Range | 0.5 mU/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction