Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ HDAC Activity Assay Kit (Fluorometric)
Click to view available options
Quantity:
1 Kit
Description
Includes
- HDAC Substrate [Boc-Lys(Ac)-AMC, 4mM], 500μL
- 10x HDAC Assay Buffer, 1mL
- Lysine Developer, 1mL
- HDAC inhibitor (Trichostatin A, 1mM), 10μL
- HeLa nuclear extract (5mg/mL), 10uL
- Deacetylated standard [Boc-Lys-AMC, 4mM], 20μL
Specifications
Specifications
Content And Storage | Store the kit at -80°C. |
Description | HDAC Activity Assay Kit (Fluorometric) is used for providing a fast and fluorescence-based method that eliminates radioactivity, extractions, or chromatography, as used in traditional assays. |
Detection Method | Fluorometric |
For Use With (Application) | Functional Study |
Includes | 500μL HDAC substrate (Boc-Lys(Ac)-AMC, 4mM), 1mL HDAC assay buffer 10X, 1mL Lysine developer, 10μL HDAC Inhibitor (Trichostatin A, 1mM), 10μL HeLa nuclear extract (5mg/mL), 20μL Deace |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Lysate, Plasma, Serum, Tissue Lysate, Urine |
Type | Activity Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction