Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ L-Alanine Assay Kit
Shop All Abnova Corporation Products
Click to view available options
Calibration Range:
Colorimetric: 1 to 200μM, Fluorimetric: 0.4 to 20μM
Description
Includes
- Alanine assay buffer, 25mL
- Alanine probe (in DMSO), 0.2mL
- Alanine converting enzyme (lyophilized), 1 vial
- Alanine development mix (lyophilized), 1 vial
- Alanine standard (10μmol, lyophilized), 1 vial
Specifications
Specifications
Calibration Range | Colorimetric: 1 to 200μM, Fluorimetric: 0.4 to 20μM |
Content And Storage | Store the kit at -20°C. |
Description | L-Alanine Assay Kit is a quantitative colorimetric/fluorimetric determination of L-alanine at OD570, FL530/585 nm. |
Detection Method | Colorimetric, Fluorometric |
For Use With (Application) | Functional Study |
Includes | Developer (6mL), ALT Enzyme (120μL), Dye Reagent (120μL), Cosubstrate (600μL), Alanine Standard (400μL ) |
Quantity | 1 Kit |
Sample Type | Culture Medium, Plasma, Serum, Tissue, Urine |
Sample Volume | 50 μL |
Type | Detection Assay Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction