Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phenylalanine Assay Kit (Fluorometric)

Catalog No. 89101197
Click to view available options
Calibration Range:
0.1 to 10nm
2 to 300μM

Includes

  • Phenylalanine Assay Buffer 25mL
  • Tyrosinase lyophilized
  • Enzyme Mix lyophilized
  • Developer lyophilized
  • Phenyalanine Standard (10.0μmol) lyophilized

To quantify PHE, functional study

Specifications

Calibration Range 2 to 300μM
Content And Storage Store the kit at -20°C.
Description Phenylalanine Assay Kit is a quantitative fluorimetric determination of phenylalanine at FL530/595 nm.
Detection Method Fluorometric
For Use With (Application) Functional Study
Includes Assay Buffer (10mL), NAD Solution (1mL), Probe (750μL), Enzyme A (1 vial), Enzyme B (120μL), Standard (120μL)
Quantity 1 Kit
Sample Type Serum, Urine
Sample Volume 10 μL
Type Phenylalanine Assay Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.