Learn More
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Agglutination, Immunofluorescence
Specifications
Specifications
| Antigen | ABO, Blood Group A Antigen |
| Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Agglutination |
| Classification | Monoclonal |
| Clone | HE-10 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml, Agglutination |
| Formulation | 10mM PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Accession No. | P16442 |
| Gene Alias | A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
