Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABO, Blood Group A Antigen Mouse anti-Human, Clone: 33C13, Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247866B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247866B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247866B Supplier Novus Biologicals Supplier No. NBP2478660.2MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

ABO, Blood Group A Antigen Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen ABO, Blood Group A Antigen
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone 33C13
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 0.5 - 1.0 ug/ml
Formulation PBS with No Preservative
Gene Alias A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Gene Symbols ABO
Host Species Mouse
Immunogen Mucin isolated from an ovarian cyst fluid
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Test Specificity This MAb preferably reacts with determinants of chain A and H type 3(Gal1-3GalNAc-R) and 4 (Gal1-3GalNAc-R), but not with type 1 and 2 chain structures. It is not reactive with immuno-dominant A trisaccharide. This MAb is applicable for tissue staining in tumor patients with blood groups A and AB. It shows a highly heterogeneous reactivity in human colon tumor tissue and adjacent mucosa. Blood-group antigens are generally defined as molecules formed by sequential addition of saccharides to the carbohydrate side chains of lipids and proteins detected on erythrocytes and certain epithelial cells. The A, B and H antigens are reported to undergo modulation during malignant cellular transformation. Blood group related antigens represent a group of carbohydrate determinants carried on both glycolipids and glycoproteins. They are usually mucin-type, and are detected on erythrocytes, certain epithelial cells, and in secretions of certain individuals. Sixteen genetically and biosynthetically distinct but inter-related specificities belong to this group of antigens, including A, B, H, Lewis A, Lewis B, Lewis X, Lewis Y, and precursor type 1 chain antigens.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.