Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABO, Blood Group H Antigen Antibody (19-OLE), Novus Biologicals™
SDP

Catalog No. NBP24462802 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24462802 0.2 mg
NBP24462800 0.02 mg
NBP24462801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24462802 Supplier Novus Biologicals Supplier No. NBP2446280.2MG

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence

ABO, Blood Group A Antigen Monoclonal antibody specifically detects ABO, Blood Group A Antigen in Human samples. It is validated for Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen ABO, Blood Group A Antigen
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone 19-OLE
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P16442
Gene Alias A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Gene Symbols ABO
Host Species Mouse
Immunogen Mucinous colonic adenocarcinoma
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.