Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ABP 688

Catalog No. p-7110792
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity human mGlu5 receptor antagonist

ABP 688 is a high affinity human mGlu5 receptor antagonist (Ki = 1.7 nM). Inhibits glutamate-induced calcium release from L(tk-) cells expressing human mGlu5 receptors (IC50 = 2.3 nM).

Specifications

Chemical Name or Material 3-[2-(6-Methyl-2-pyridinyl)ethynyl]-2-cyclohexene-1-one O-methyloxime
CAS 924298-51-1
Quantity 10 mg
Target Group I mGluR Antagonists
Molecular Formula C15H16N2O
Formula Weight 240.3
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.