Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABP1/AOC1 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB091374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB091374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB091374 Supplier Novus Biologicals Supplier No. NBP300162AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ABP1/AOC1 Polyclonal specifically detects ABP1/AOC1 in Human samples. It is validated for ELISA.

Specifications

Antigen ABP1/AOC1
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias ABP, amiloride binding protein 1 (amine oxidase (copper-containing)), Amiloride-binding protein, amiloride-sensitive amine oxidase, amiloride-sensitive amine oxidase [copper-containing], AOC1DAOamiloride-binding protein-1, DAO1, Diamine oxidase, EC 1.4.3, EC 1.4.3.22, Histaminase, KAO, Kidney amine oxidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ABP1/AOC1 (AAH14093.1; Met1-Val751).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.