Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ABpure™ Antibody Purification Kit
SDP

Catalog No. p-7111140 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
26-000-05 1 Purification
26-000-10 3 Purifications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-000-05 Supplier Novus Biologicals™ Supplier No. 2600005

Novus Biologicals™ ABpure Antibody Purification Kit is used to remove contaminants such as BSA and other proteins, glycine, tris, and azide that interfere in conjugation reactions.

  • Used to purify antibodies from crude samples such as ascites fluid or immune serum
  • Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
  • 20 to 500μg of antibody can be purified in each run

Specifications

Content And Storage 4°C
Quantity 1 Test
Kit Contents 4 or 12 additional collecting tubes, 1 vial AbPure wash buffer, 1 vial AbPure eution buffer, 1 vial AbPure neutralization buffer, 1 or 3 vials Protein A resin, 1 vial 10X AbPure binding buffer, 1/3 spin cartridge/collecting tube assemblies
Sufficient For 1 Purification
Product Type Antibody Purification Kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.