Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Novus Biologicals™ AbPure™ BSA Removal Antibody Purification Kit Catalog No.: 2630100

Novus Biologicals™ AbPure™ BSA Removal Antibody Purification Kit
SDP

Catalog No. 2630100
1 mL
Click to view available options
Quantity:
1 mL

Catalog No. 26-301-00

Supplier: Novus Biologicals™ 2630100

Novus Biologicals™ AbPure BSA Removal Kit can be used on any antibody sub-type, and species.

Kit Components:
  • 1 x vial AbPure BSA Removal Buffer
  • 1 x vial AbPure Re-Suspension Buffer

Specifications

Content And Storage 4°C
Quantity 1 mL
Kit Contents 1 x vial AbPure BSA Removal Buffer, 1 x vial AbPure Re-Suspension Buffer
Product Type AbPure BSA Removal Kit

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ AbPure™ BSA Removal Antibody Purification Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.