Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, fluorescein
Incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.
Specifications
Concentration | 2X |
---|---|
Detection Method | SYBR |
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Description
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with ROX or Fluorescein passive reference dyes
Specifications
2X | |
Tube | |
qPCR | |
Standard | |
ABsolute Blue | |
Gene Expression, Genotyping, siRNA Validation | |
DNA, dsDNA |
SYBR | |
High | |
Taq DNA Polymerase | |
BioRad iCycler iQ, BioRad MyiQ, BioRad iQ5 | |
Real Time PCR SYBR Master Mix | |
qPCR Cyclers |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.