Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial
Description
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with ROX or Fluorescein passive reference dyes
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Contains: • 16 x 1.25 mL ABsolute Blue QPCR SYBR Green Mix (2X) • 100 μL MgCl2 (1M) • 25 μL ROX Reference Dye (1 mM) Sufficient for 1600 x 25 μL reactions. Store at -20°C |
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene |
| Product Line | ABsolute Blue |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.