Learn More
Thermo Scientific™ ABsolute QPCR Mix, low ROX
Optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes.
$1066.00 - $3760.00
Specifications
Concentration | 2X |
---|---|
Detection Method | Primer-probe |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Description
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.
- High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
- Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
- Optimized buffer - For maximal performance in multiplex reactions
Specifications
2X | |
High | |
Taq DNA Polymerase | |
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P | |
qPCR Mix | |
2 X |
Primer-probe | |
qPCR | |
Standard | |
ABsolute | |
Gene Expression | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.