Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ABT 199
GREENER_CHOICE

Catalog No. p-7166834
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective, high affinity Bcl-2 inhibitor; orally bioavailable

ABT 199 is a selective, high affinity Bcl-2 inhibitor (Ki < 0.010 nM). Exhibits >4800-fold selectivity for Bcl-2 over Bcl-xL and Bcl-w, and displays no measurable activity at Mcl-1 (Ki > 444 nM). Potently induces apoptosis in FL5.12-BCL-2 cells (EC50 = 261 nM) and reduces tumor burden in chronic lymphocytic leukemia (CLL) primary samples (EC50 = 3 nM). Shows reduced toxicity to platelets compared to similar compounds. Enhances efficacy of clinically relevant chemotherapy and immunotherapy drugs. Orally bioavailable. Exhibits binding to SARS-CoV-2 3C-like protease (3CLpro) active site in a virtual screen. ABT 199 reverses oxidative phosphorylation in acute myeloid leukemia cells (AML), in vitro and in vivo.

Specifications

Chemical Name or Material 4-[4-[[2-(4-Chlorophenyl)-4,4-dimethyl-1-cyclohexen-1-yl]methyl]-1-piperazinyl]-N-[[3-nitro-4-[[(tetrahydro-2 H-pyran-4-yl)methyl]amino]phenyl]sulfonyl]-2-(1 H-pyrrolo[2,3-b]pyridin-5-yloxy)benzamide
CAS 1257044-40-8
Synonym GDC-0199
Quantity 10 mg
Target Bcl-2 Family Inhibitors
Molecular Formula C45H50ClN7O7S
Formula Weight 868.45
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.