Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ABT 737
GREENER_CHOICE

Catalog No. p-7166804
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Bcl-2 family inhibitor; mimics BH3 and induces apoptosis in cancer cell lines

ABT 737 is a high affinity Bcl-2 family inhibitor (Ki ≤ 1nM for Bcl-XL, Bcl-2 and Bcl-w). BH3 mimetic; triggers Bax/Bak-mediated apoptosis. Exhibits cytotoxicity in lymphoma cell lines overexpressing bcl-2 (EC50 < 1 μM) and multiple myeloma cells (IC50 = 5-15 μM). Induces apoptosis in patient-derived CLL cells. Promotes regression of SCLC tumors in a mouse xenograft model. Synergizes with certain other anticancer agents.

Specifications

Quantity 10 mg
Molecular Formula C42H45ClN6O5S2
Formula Weight 813.43
Product Type Inhibitor
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.