Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ABX 464
GREENER_CHOICE

Catalog No. p-7250156
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

HIV RNA splicing modulator

ABX 464 is an HIV RNA splicing modulator; it selectively enhances the splicing of HIV RNA in infected human PBMCs and inhibits viral replication, with no global effect on cellular splicing. ABX 464 inhibits replication of HIV strains harboring mutations associated with drug-resistance. ABX 464 binds the cap binding complex (CBC) and upregulates miR-124 expression in vitro and decreases expression of miR-124 target genes, MCP-1, CXCL-1, SERPIN-E1 in macrophages. It also decreases the number of Th17 cells, as well as IL-6 soluble receptor in CD4+ T cells. In HIV infected humanized mice, treatment with ABX 464 reduces viral load. Anti-inflammatory.

Specifications

Chemical Name or Material 8-Chloro-N-[4-(trifluoromethoxy)phenyl]-2-quinolinamine
CAS 1258453-75-6
Quantity 10 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C16H10ClF3N2O
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.