Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Acacia
SCI_ED

Catalog No. s25114 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
Packaging:
Solid

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 9000-01-5
Synonym Gum Arabic

Specifications

Assay Percent Range >99%
Density 1.4 to 1.5g/mL
Quantity 100 g
Packaging Solid
Chemical Name or Material Acacia
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.