Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAD9 Antibody, Novus Biologicals™
SDP

Catalog No. NB433343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB433343 25 μL
NBP182749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB433343 Supplier Novus Biologicals Supplier No. NBP18274925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACAD9 Polyclonal specifically detects ACAD9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen ACAD9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50, Knockdown Validated
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9H845
Gene Alias ACAD-9, acyl-CoA dehydrogenase family member 9, mitochondrial, acyl-CoA dehydrogenase family, member 9, acyl-Coenzyme A dehydrogenase family, member 9, EC 1.3.99, EC 1.3.99.-, EC 1.3.99.10, FLJ23533, MGC14452, NPD002
Gene Symbols ACAD9
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:EILRMYIALTGLQHAGRILTTRIHELKQAKVSTVMDTVGRRLRDSLGRTVDLGLTGNHGVVHPSLADSANKFEENTYCFGRTVETLL
Molecular Weight of Antigen 69 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28976
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.