Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACADSB Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB320139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320139 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320139 Supplier Novus Biologicals Supplier No. NBP298594APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACADSB Polyclonal antibody specifically detects ACADSB in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen ACADSB
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 2-MEBCAD, 2-methylbutyryl-coenzyme A dehydrogenase, ACAD7,2-methylbutyryl-CoA dehydrogenase, acyl-CoA dehydrogenase, short/branched chain, acyl-Coenzyme A dehydrogenase, short/branched chain, EC 1.3.99,2-methyl branched chain acyl-CoA dehydrogenase, EC 1.3.99.-, SBCADshort/branched chain specific acyl-CoA dehydrogenase, mitochondrial
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ACADSB fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 36
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.