Learn More
Description
Specifications
Specifications
| Antigen | ACADSB |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | 2-MEBCAD, 2-methylbutyryl-coenzyme A dehydrogenase, ACAD7,2-methylbutyryl-CoA dehydrogenase, acyl-CoA dehydrogenase, short/branched chain, acyl-Coenzyme A dehydrogenase, short/branched chain, EC 1.3.99,2-methyl branched chain acyl-CoA dehydrogenase, EC 1.3.99.-, SBCADshort/branched chain specific acyl-CoA dehydrogenase, mitochondrial |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ACADSB fragment. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
