Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7205535 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9166 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9166 Supplier Proteintech Group Inc Supplier No. 140291AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation-the process of receptor-mediated endocytosis. Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits. It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells.

Specifications

Antigen ACAP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ACAP2
Gene Accession No. Q15057, Q5FVC7, Q6ZQK5
Gene Alias ACAP2, Centaurin beta 2, CENTB2, CNT B2, KIAA0041
Gene Symbols Acap2
Host Species Rabbit
Immunogen ACAP2 Fusion Protein Ag5190
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23527, 619382, 78618
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.