Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACAT Antibody, Novus Biologicals™
SDP

Catalog No. NB400141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400141 0.1 mL
NB400141SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400141 Supplier Novus Biologicals Supplier No. NB400141
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

ACAT Polyclonal specifically detects ACAT in Human, Mouse, Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen ACAT
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunoprecipitation 2-10 ug/ml lysate
Formulation Whole antisera with 0.02% Sodium Azide
Gene Accession No. P35610
Gene Alias ACACT, ACACT1, ACAT-1, ACATACAT1, acyl-Coenzyme A: cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase 1, Cholesterol acyltransferase 1, EC 2.3.1.26, RP11-215I23.2, STATSOAT, sterol O-acyltransferase (acyl-Coenzyme A: cholesterol acyltransferase) 1, sterol O-acyltransferase 1
Gene Symbols SOAT1
Host Species Rabbit
Immunogen A fusion protein construct that contains the N-terminal region of human ACAT1. [UniProt# P35610]
Molecular Weight of Antigen 46 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6646
Test Specificity This product is specific for protein corresponding to ACAT1 - Sterol O-acyltransferase 1 (SOAT1) Uniprot# P35610, Gene ID# 6646. [Important Note:- ACAT1 is a symbol for both acyl-Coenzyme A: cholesterol acyltransferase 1 (SOAT1/ACAT1) and for acetyl-CoA acetyltransferase 1 (ACAT1/ THIL, Gene ID 38, Uniprot P24752) which often cause confusion].
Target Species Human, Mouse, Hamster
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.