Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACAT1 (CES1) Mouse anti-Human,Monkey, Unconjugated, Clone: 10E8, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | ACAT1 (CES1) |
---|---|
Clone | 10E8 |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC11012MI
|
MilliporeSigma
MABC110125UG |
25 μg |
Each for $189.29
|
|
|||||
MABC11011MI
|
MilliporeSigma
MABC1101100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
ACAT1 (CES1) Monoclonal antibody specifically detects ACAT1 (CES1) in Human,Monkey samples. It is validated for ELISA, Western Blotting.Specifications
ACAT1 (CES1) | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human, Monkey | |
Liver carboxylesterase 1;EC: 3.1.1.13;Acyl-coenzyme A:cholesterol acyltransferase;ACAT;Brain carboxylesterase hBr1;Carboxylesterase 1;CE-1;hCE-1;Cocaine carboxylesterase;Egasyn;HMSE;Monocyte/macrophage serine esterase;Retinyl ester hydrolase;REH;Serine esterase 1;Triacylglycerol hydrolase;TGH | |
A linear peptide corresponding to 14 amino acids from the C-terminal region of human Liver Carboxyesterase 1 (ACAT1). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼62 kDa observed; 62.52 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
10E8 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
NP_001020365 | |
CES1, CES2, SES1 | |
IgG1 κ | |
Protein G Purified | |
Clone 10E8 is a mouse monoclonal antibody that specifically detects human liver carboxyesterase 1 (ACAT1). It targets an epitope with in 14 amino acids from the C-terminal region. It is expected to react with all isoforms of ACAT1. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title