Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACBD3 Antibody (OTI3A1), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP272149R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272149R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272149R Supplier Novus Biologicals Supplier No. NBP272149R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACBD3 Monoclonal specifically detects ACBD3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen ACBD3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3A1
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias acyl-CoA binding domain containing 3, Acyl-CoA-binding domain-containing protein 3, acyl-Coenzyme A binding domain containing 3, GCP60Golgi phosphoprotein 1, GOCAP1Peripheral benzodiazepine receptor-associated protein PAP7, golgi complex associated protein 1, 60kDa, Golgi complex-associated protein 1, Golgi resident protein GCP60, GOLPH1PBR- and PKA-associated protein 7, PAP7, PBR associated protein, peripherial benzodiazepine receptor associated protein, PKA (RIalpha)-associated protein
Gene Symbols ACBD3
Host Species Mouse
Immunogen Full length recombinant protein of human ACBD3 (NP_073572) produced in HKE293T cell.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 64746.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.