Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACBD6 Antibody (4A9D5E9), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB351066 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351066 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351066 Supplier Novus Biologicals Supplier No. NBP306614MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ACBD6 Monoclonal antibody specifically detects ACBD6 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen ACBD6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone 4A9D5E9
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ACBD6 (Accession#: NP_115736.1; Met 1-Ala 282). .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84320
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.