Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACBP Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB080643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB080643 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB080643 Supplier Novus Biologicals Supplier No. NBP297012AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ACBP Polyclonal specifically detects ACBP in Human samples. It is validated for ELISA.

Specifications

Antigen ACBP
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias acyl-Coenzyme A binding domain containing 1, acyl-Coenzyme A bindingprotein), CCK-RP, cholecystokinin-releasing peptide, trypsin-sensitive, diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein), Diazepam-binding inhibitor, endozepine, EP, GABA receptor modulator, MGC70414
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ACBP (P07108-2; Met1-Ala87).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1622
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.