Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Accessories for Microplate Reader
GSA_VAGREENER_CHOICE

Catalog No. p-4672017 Shop All Agilent Technologies Inc. Products
Click to view available options
Product Type:
Fluorescence Filter
This item is not returnable. View return policy

Catalog No. BT7082225

Supplier: Agilent BioTek 7082225

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with Multi-mode, absorbance and monochromator based microplate readers.

Serial Cable (BT75034)

  • Modem (9-pin female to 9-pin male)
  • For use with ELx800, Synergy 2, Synergy 2 Alpha, Synergy 2 SL, Synergy H1m, Synergy HT, Synergy Mx, Synergy MxF
Serial Cable (BT75053)
  • Null Modem (9-pin female to 25-pin female)
  • For use with ELx808, FLx800, PowerWave, Precision, Precision XS, Synergy HT
Fluorescence Filter (BT7082225)
  • 590/20 nm
  • For use with Cytation3, FLx800, Synergy 2, Synergy H1, Synergy H4, Synergy HT
Fuse (BT46024)
  • 250V 1.5A
  • For use with ELx808
Fuse (BT46023)
  • 0.75A
  • For use with PowerWave X

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Fluorescence Filter
For Use With (Equipment) Agilent BioTek Microplate Readers
Excitation/Emission 590/20 nm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.