Learn More
Cytiva Accessories for Multiphor™ II Electrophoresis System
Catalog No. 45000423 Shop All Cytiva Products
Change view
Click to view available options
Type:
EPH Electrode Anode
EPH Electrode Wick (104 x 253 mm)
EPH Electrode Wick (82 x 130 mm)
Flexiclamp
IEF Electrode Strip
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
45-000-423 Flexiclamp
45-000-418 IEF Electrode Strip
45-000-461 EPH Electrode Anode
09-928-082 EPH Electrode Wick (82 x 130 mm)
45-001-627 EPH Electrode Wick (104 x 253 mm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 45-000-423 Supplier Cytiva Supplier No. 18101373
Specifications
For Use With (Application) SDS-PAGE/IEF standard flatbed unit
For Use With (Equipment) Multiphor II Electrophoresis System
Type Flexiclamp